Learn More
Kingfisher Biotech Inc Bovine VEGF-A ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1906B004
Encompass_Preferred
The Bovine VEGF-A ELISA contains capture antibody, recombinant protein standard, and detection antibody for development of a Bovine VEGF-A ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Bovine VEGF-A ELISA can also be used to measure Bison, and Water Buffalo VEGF-A. Alternate names include VEGFA, MVCD1, VEGF, VPF, vascular endothelial growth factor A. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5588