Learn More
Biomatik Corporation Bovine Cystatin C (Cys-C) ELISA Kit
Supplier:  Biomatik Corporation EKU10768
Encompass_Preferred
Bovine Cystatin C (Cys-C) ELISA Kit for detecting Cys-C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 27.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01035. Gene ID: 281102. Target Synonyms: CST3; Cystatin 3; Gamma Trace; Post-Gamma-Globulin; Neuroendocrine Basic Polypeptide; Amyloid Angiopathy And Cerebral Hemorrhage. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7461