Learn More
Raybiotech Inc C-Series Bovine Cytokine Antibody Array 1 Kit.
Catalog No. 501948448
Encompass_Preferred
Detects 30 Bovine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: aFGF, Angiopoietin-1, bFGF, CD40 Ligand, Decorin, GASP-1, IFN alpha, IFN-beta, IFN-gamma, IGF-1, IL-1 alpha, IL-1 beta, IL-1 F5, IL-10, IL-13, IL-15, IL-17A, IL-18, IL-2, IL-21, IL-4, IP-10, LIF, MCP-1, MIG, MIP-1 beta, NCAM-1, RANTES, TNF alpha, VEGF-A
Catalog No. 50-194-8448 Supplier Raybiotech Inc Supplier No. AABCYT12
Detects 30 Bovine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: aFGF, Angiopoietin-1, bFGF, CD40 Ligand, Decorin, GASP-1, IFN alpha, IFN-beta, IFN-gamma, IGF-1, IL-1 alpha, IL-1 beta, IL-1 F5, IL-10, IL-13, IL-15, IL-17A, IL-18, IL-2, IL-21, IL-4, IP-10, LIF, MCP-1, MIG, MIP-1 beta, NCAM-1, RANTES, TNF alpha, VEGF-A