Learn More
Raybiotech Inc G-Series Bovine Cytokine Array 1 Kit.
Catalog No. 501950330
Encompass_Preferred
Semi-quantitative measurement of 10 bovine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IFN alpha, IFN-gamma, IL-13, IL-1 alpha (IL-1 F1), IL-1 F5 (FIL1 delta), IL-21, IP-10 (CXCL10), MIG (CXCL9), MIP-1 beta (CCL4), TNF alpha
Catalog No. 50-195-0330 Supplier Raybiotech Inc Supplier No. GSBCYT11
Semi-quantitative measurement of 10 bovine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IFN alpha, IFN-gamma, IL-13, IL-1 alpha (IL-1 F1), IL-1 F5 (FIL1 delta), IL-21, IP-10 (CXCL10), MIG (CXCL9), MIP-1 beta (CCL4), TNF alpha