Learn More
Raybiotech Inc G-Series Bovine Cytokine Array 2 Kit.
Catalog No. 501950333
Encompass_Preferred
Semi-quantitative measurement of 10 Bovine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: aFGF (FGF-1), bFGF, GASP-1 (WFIKKNRP), IGF-1, IL-2, IL-4, IL-15, MCP-1 (CCL2), NCAM-1 (CD56), VEGF-A
Catalog No. 50-195-0333 Supplier Raybiotech Inc Supplier No. GSBCYT22
Semi-quantitative measurement of 10 Bovine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: aFGF (FGF-1), bFGF, GASP-1 (WFIKKNRP), IGF-1, IL-2, IL-4, IL-15, MCP-1 (CCL2), NCAM-1 (CD56), VEGF-A