Learn More
Raybiotech Inc G-Series Bovine Cytokine Array 3 Kit.
Supplier:  Raybiotech Inc GSBCYT31
Encompass_Preferred
Semi-quantitative measurement of 10 Bovine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Angiopoietin-1, CD40 Ligand (TNFSF5), Decorin, IFN-beta, IL-1 beta (IL-1 F2), IL-10, IL-17A, IL-18, LIF, RANTES (CCL5)
Catalog No. 50-195-0334