Learn More
Raybiotech Inc Quantibody Bovine Cytokine Antibody Array 30 Kit.
Supplier:  Raybiotech Inc QABCAA302
Encompass_Preferred
A combination of 3 non-overlapping arrays to quantitatively measure 30 Bovine cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: aFGF, Angiopoietin-1, bFGF, CD40 Ligand, Decorin, GASP-1, IFN alpha, IFN-beta, IFN-gamma, IGF-1, IL-1 alpha, IL-1 beta, IL-10, IL-13, IL-15, IL-17A, IL-18, IL-2, IL-21, IL-36 Ra, IL-4, IP-10, LIF, MCP-1, MIG, MIP-1 beta, NCAM-1, RANTES, TNF alpha, VEGF-A
Catalog No. 50-195-0781