Learn More
Biomatik Corporation Bovine Defensin Beta 1 (DEFb1) ELISA Kit
Supplier:  Biomatik Corporation EKU03655
Encompass_Preferred
Bovine Defensin Beta 1 (DEFb1) ELISA Kit for detecting DEFb1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P46159. Target Synonyms: B-DF1; DEFB1; BD1; DEF-B1; DEFB101; HBD1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7247