Learn More
American Research Products Inc BoVine DeoXyribonuclease 1 DN
Catalog No. 5000328166
Encompass
BoVine DeoXyribonuclease-1 (DNASE1) ELISA kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 1 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28166 Supplier American Research Products Inc Supplier No. CSBEL007049BO
BoVine DeoXyribonuclease-1 (DNASE1) ELISA kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 1 pg/mL Sample serum plasma tissue homogenates Method Sandwich