Learn More
Biomatik Corporation Bovine Elastin (ELN) CLIA Kit
Supplier:  Biomatik Corporation EKU12427
Encompass_Preferred
Bovine Elastin (ELN) CLIA Kit for detecting ELN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.27-200ng/ml. Sensitivity: 0.09ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04985. Target Synonyms: WBS, WS, SVAS; Tropoelastin; Supravalvular Aortic Stenosis; Williams-Beuren Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7646