Learn More
Biomatik Corporation Bovine Endocrine Gland Derived Vascular Endothelial Growth Factor (EG-VEGF) CLIA Kit
Supplier:  Biomatik Corporation EKU09604
Encompass_Preferred
Bovine Endocrine Gland Derived Vascular Endothelial Growth Factor (EG-VEGF) CLIA Kit for detecting EG-VEGF in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.56pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: F1MN36. Gene ID: 617599. Target Synonyms: PK1; EGVEGF; PRK1, PROK1; Prokineticin 1; Black Mamba Toxin-Related Protein; Mambakine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6098