Learn More
Biomatik Corporation Bovine Eosinophil Chemotactic Factor (ECF) CLIA Kit
Supplier:  Biomatik Corporation EKU09608
Encompass_Preferred
Bovine Eosinophil Chemotactic Factor (ECF) CLIA Kit for detecting ECF in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 2.48pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9TTS6. Gene ID: 404072. Target Synonyms: CCL11; SCYA11; Eotaxin 1; Eosinophil Chemotactic Protein 1; Chemokine C-C-Motif Ligand 11; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8485