Learn More
American Research Products Inc BoVine Epithelial growth facto
Catalog No. 5000328441
Encompass
BoVine Epithelial growth factor (EGF) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28441 Supplier American Research Products Inc Supplier No. CSBE17171B
BoVine Epithelial growth factor (EGF) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma Method Sandwich