Learn More
Biomatik Corporation Bovine Epithelial Growth Factor (Egf) Elisa Kit
Supplier:  Biomatik Corporation EKC31624
Encompass_Preferred
Bovine Epithelial Growth Factor (Egf) Elisa Kit for detecting Epithelial Growth Factor(Egf) in serum, plasma. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: HOMG4; URG; beta-urogastrone; epidermal growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6634