Learn More
Kingfisher Biotech Inc Bovine EPO Polyclonal Antibody (100 ug)
SDP
Supplier:  Kingfisher Biotech Inc KP1583B100
Encompass
The Bovine Erythropoietin (EPO) polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Bovine Erythropoietin ELISA. The Bovine Erythropoietin antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Bovine EPO - Strong) (Canine EPO - Strong) (Equine EPO - Strong) (Feline EPO - Strong) (Mouse EPO - Strong) (Rabbit EPO - Strong) (Swine EPO - Strong). For research use only. Made in the USA
Catalog No. 50-233-2438