Learn More
Enzyme Research Laboratories Inc BOVINE FACTOR XA 1MG BOVINE FA
Catalog No. NC0156622
Encompass
Bovine Factor Xa is prepared from BFX by activation with Russell's Viper Venom. Complete activation is observed by SDS-PAGE.
Catalog No. NC0156622 Supplier Enzyme Research Laboratories Inc Supplier No. BFXA
Bovine Factor Xa is prepared from BFX by activation with Russell's Viper Venom. Complete activation is observed by SDS-PAGE.