Learn More
R&D Systems™ Bovine FGF acidic/FGF1 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 281160 |
| Name | FGF acidic |
| Quantity | 25 μg |
| Source | Bovine brain tissue-derived FGF acidic/FGF1 protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.