Learn More
Gibco™ Bovine FGF-basic (FGF-2/bFGF), Animal-Free Recombinant Protein, PeproTech®
Description
Recombinant Bovine FGF-basic is a 17.1 kDa protein consisting of 154 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | P03969 |
| For Use With (Application) | Control, Bioactivity |
| Format | Lyophilized |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 281161 |
| Molecular Weight (g/mol) | 17.1 kDa |
| Name | Bovine FGF-basic (FGF-2/bFGF), Animal-Free |
| Purification Method | Purified |
| Quantity | 250 μg |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.