Learn More
Biomatik Corporation Bovine Fibroblast Growth Factor 4 (FGF4) CLIA Kit
Supplier:  Biomatik Corporation EKU09716
Encompass_Preferred
Bovine Fibroblast Growth Factor 4 (FGF4) CLIA Kit for detecting FGF4 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.22pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P48803. Target Synonyms: HBGF4; HST; HSTF1; KFGF; K-FGF; Heparin Secretory Transforming Protein 1; Kaposi Sarcoma Oncogene; Transforming Protein KS3; FGF-Related Oncogene. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8458