Learn More
American Research Products Inc BoVine Fibroblast Growth Facto
Catalog No. 5000328429
Encompass
BoVine Fibroblast Growth Factor 23 (FGF23) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28429 Supplier American Research Products Inc Supplier No. CSBE16193B
BoVine Fibroblast Growth Factor 23 (FGF23) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich