Learn More
American Research Products Inc BoVine fibroblast growth facto
Catalog No. 5000326922
Encompass
BoVine fibroblast growth factor 21 (FGF21) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26922 Supplier American Research Products Inc Supplier No. CSBEL008627BO
BoVine fibroblast growth factor 21 (FGF21) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich