Learn More
Raybiotech Inc Bovine Fibronectin ELISA Kit
Catalog No. 501948858
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 1.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Bovine Fibronectin (FN1)
Catalog No. 50-194-8858 Supplier Raybiotech Inc Supplier No. ELBFN11
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 1.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Bovine Fibronectin (FN1)