Learn More
American Research Products Inc BoVine follicle stimulating ho
Catalog No. 5000330142
Encompass
BoVine follicle-stimulating hormone (FSH) ELISA Kit from CUSABIO Detection Range 2 mIU/mL-800 mIU/mL Sensitivity 2 mIU/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-30142 Supplier American Research Products Inc Supplier No. CSBE15856B
BoVine follicle-stimulating hormone (FSH) ELISA Kit from CUSABIO Detection Range 2 mIU/mL-800 mIU/mL Sensitivity 2 mIU/mL Sample serum plasma tissue homogenates Method Competitive