Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Bovine Foot (Bos taurus) Specimen
SDPSCI_ED

Catalog No. S00568 Shop All 3B Scientific Products
Change view
Click to view available options
For Use With (Application):
Comparative Biology
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S00568 Comparative Biology
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00568 Supplier 3B Scientific™ Supplier No. 1021063
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3B Scientific Bovine Foot Specimen can be used for comparative anatomy.

  • Bovine foot (Bos taurus)
  • Specimen
  • For use in comparative biology

Specifications

Color White, Brown
For Use With (Application) Comparative Biology
Weight (English) 3.97 lb.
Product Type Bovine Foot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.