Learn More
American Research Products Inc BoVine free tri iodothyronine
Catalog No. 5000328887
Encompass
BoVine free tri-iodothyronine (FT3) ELISA kit from CUSABIO Detection Range 2 pmol/L-32 pmol/L Sensitivity 0 38 pmol/L Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-28887 Supplier American Research Products Inc Supplier No. CSBEQF027510BO
BoVine free tri-iodothyronine (FT3) ELISA kit from CUSABIO Detection Range 2 pmol/L-32 pmol/L Sensitivity 0 38 pmol/L Sample serum plasma cell culture supernates tissue homogenates Method Competitive