Learn More
Carbosynth LLC. Purified Bovine Gamma Globulin (99% pure)
SDP
Catalog No. 501252964
Encompass_Preferred
Gamma globulins are a class of globulins, identified by their position after serum protein electrophoresis. The most significant gamma globulins are immunoglobulins ("Igs"), more commonly known as antibodies, although some Igs are not gamma globulins, and some gamma globulins are not Igs.
Catalog No. 50-125-2964 Supplier Carbosynth LLC. Supplier No. 30AB4410G
Gamma globulins are a class of globulins, identified by their position after serum protein electrophoresis. The most significant gamma globulins are immunoglobulins ("Igs"), more commonly known as antibodies, although some Igs are not gamma globulins, and some gamma globulins are not Igs.