Learn More
Biomatik Corporation Bovine Gelsolin (GSN) ELISA Kit
Supplier:  Biomatik Corporation EKU10862
Encompass_Preferred
Bovine Gelsolin (GSN) ELISA Kit for detecting GSN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.6ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3SX14. Gene ID: 535077. Target Synonyms: GS; AGEL; ADF; Brevin; Gelsolin Amyloidosis,Finnish Type; Actin-depolymerizing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5681