Learn More
American Research Products Inc BoVine gonadotropin releasing
Catalog No. 5000328591
Encompass
BoVine gonadotropin-releasing hormone (GnRH) ELISA Kit from CUSABIO Detection Range 100 pg/mL-4000 pg/mL Sensitivity 100 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28591 Supplier American Research Products Inc Supplier No. CSBE15857B
BoVine gonadotropin-releasing hormone (GnRH) ELISA Kit from CUSABIO Detection Range 100 pg/mL-4000 pg/mL Sensitivity 100 pg/mL Sample serum plasma tissue homogenates Method Competitive