Learn More
American Research Products Inc BoVine Growth differentiation
Catalog No. 5000327112
Encompass
BoVine Growth/differentiation factor 8 (MSTN) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27112 Supplier American Research Products Inc Supplier No. CSBEL015057BO
BoVine Growth/differentiation factor 8 (MSTN) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich