Learn More
American Research Products Inc BoVine Heat Shock Protein 70
Catalog No. 5000329429
Encompass
BoVine Heat Shock Protein 70 HSP-70 ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-20 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29429 Supplier American Research Products Inc Supplier No. CSBE13452B
BoVine Heat Shock Protein 70 HSP-70 ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-20 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma tissue homogenates Method Competitive