Learn More
American Research Products Inc BoVine Heat Shock Protein 72
Catalog No. 5000328587
Encompass
BoVine Heat Shock Protein 72 (HSP-72) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28587 Supplier American Research Products Inc Supplier No. CSBE15831B
BoVine Heat Shock Protein 72 (HSP-72) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich