Learn More
American Research Products Inc BoVine Heat shock protein HSP
Catalog No. 5000326947
Encompass
BoVine Heat shock protein HSP 90-alpha (HSP90AA1) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26947 Supplier American Research Products Inc Supplier No. CSBEL010802BO
BoVine Heat shock protein HSP 90-alpha (HSP90AA1) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich