Learn More
Biomatik Corporation Bovine Hemoglobin (HB) ELISA Kit
Supplier:  Biomatik Corporation EKU08724
Encompass_Preferred
Bovine Hemoglobin (HB) ELISA Kit for detecting HB in serum, plasma, tissue homogenates, erythrocyte lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 12.35-1,000ug/ml. Sensitivity: 4.31ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01966; P02070. Gene ID: 100140149; 280813. Target Synonyms: Hgb; Haemoglobin; Heterotetramer (αβ) 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4860