Learn More
American Research Products Inc BoVine Heparin binding growth
Catalog No. 5000327299
Encompass
BoVine Heparin-binding growth factor 2 (FGF2) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27299 Supplier American Research Products Inc Supplier No. CSBEL008625BO
BoVine Heparin-binding growth factor 2 (FGF2) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich