Learn More
Kingfisher Biotech Inc Bovine IFN alpha A Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0008B025
Encompass
The Bovine IFN alpha A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IFN alpha A applications are for cell culture, ELISA standard, and Western Blot Control. The Bovine IFN alpha A yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IFN alpha A Specifications: (Molecular Weight: 19.0 kDa) (Amino Acid Sequence: CHLPHTHSLA NRRVLMLLQQ LRRVSPSSCL QDRNDFEFLQ EALGGSQLQK AQAISVLHEV TQHTFQLFST EGSPATWDKS LLDKLRAALD QQLTDLQACL TQEEGLRGAP LLKEDSSLAV RKYFHRLTLY LQEKRHSPCA WEVVRAEVMR AFSSSTNLQE SFRRKD (166)) (Gene ID: 515951). For research use only. Made in the USA
Catalog No. 50-241-4013