Learn More
Mabtech Inc Bovine IFN-gamma ELISpot BASIC kit (ALP)
Supplier:  Mabtech Inc 31192A
Encompass
The bovine IFN-gamma ELISpot kit is ideal for users who want the flexibility of setting up their own assay. The kit enables enumeration of cells secreting bovine IFN-gamma, and contains a matched pair of monoclonal capture and detection antibodies and Streptavidin-ALP.
Catalog No. NC0394904