Learn More
Kingfisher Biotech Inc Bovine IFN gamma Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0013B025
Encompass
The Bovine IFN gamma yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IFN gamma applications are for cell culture, ELISA standard, and Western Blot Control. The Bovine IFN gamma yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IFN gamma Specifications: (Molecular Weight: 16.9 kDa) (Amino Acid Sequence: QGQFFREIEN LKEYFNASSP DVAKGGPLFS EILKNWKDES DKKIIQSQIV SFYFKLFENL KDNQVIQRSM DIIKQDMFQK FLNGSSEKLE DFKKLIQIPV DDLQIQRKAI NELIKVMNDL SPKSNLRKRK RSQNLFRGRR AST) (Gene ID: 281237). For research use only. Made in the USA
Catalog No. 50-241-3957