Learn More
Raybiotech Inc Bovine IFN-gamma ELISA Kit
Supplier:  Raybiotech Inc ELBIFNG1
Encompass_Preferred
For cell culture supernatants, EDTA and/or Citrate treated plasma, and serum samples. Heparin is not recommended as an anticoagulant For use in this assay. Sensitivity: 0.11 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects bovine IFN-gamma. Other species not determined.
Catalog No. 50-194-8860