Learn More
Bethyl Laboratories, Inc Bovine IgA ELISA Kit, 96-well strip plate, Sandwich ELISA, Colorimetric detection at 450 nm, Research use only, Sample types: serum, plasma, milk, colostrum
Supplier:  Bethyl Laboratories, Inc E11131
Encompass_Preferred
Bovine IgA ELISA Kit, 96-well strip plate, Sandwich ELISA, Colorimetric detection at 450 nm, Research use only, Sample types: serum, plasma, milk, colostrum
Catalog No. NC1727309