Learn More
Raybiotech Inc Bovine IGF-1 ELISA Kit
Supplier:  Raybiotech Inc ELBIGF11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.23 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Bovine IGF-1.
Catalog No. 50-194-8861