Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine IgG

Catalog No. 08641402 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Type:
Bovine IgG
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
08641402 Bovine IgG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08641402 Supplier MP Biomedicals™ Supplier No. 08641402

missing translation for 'validMainframeMsg'

Each vial contains 100 mg of lyophilized Bovine IgG. The product was in 0.01M PBS, pH 7.4.

  • This product is useful for structural and biological studies, as a reference standard, or as starting material for immunogen preparation.
  • It is prepared using chromatographic technique.
  • Purity: >95%, Lyophilized powder.
  • Research Use Only (RUO).
  • Ready for CE IVD labeling of clinical applications.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Type Bovine IgG
pH 4.0 to 5.0
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.