Learn More
Bioworld Bovine IgG, Lyophilized Powder ≥97%, 10 g
Supplier:  Bioworld 220700262
Encompass_Preferred
Bovine Immunoglobulin G Lyophilized Powder The immunoglobulin preparations are supplied as a lyophilized powder. Purity levels are >97% IgG as analyzed by SDS-Page. These preparations are especially useful as a co-precipitant in RIA methods employing polyethylene glycol or ammonium sulfate precipitation steps, or as a protein stabilizer or blocker in a number of immunoassays. These preparations are also useful as IgG control solutions for biochemical analytical tests. Immunoglobulin preparations reconstitute to pH 7.2 ± 0.2. Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.
Catalog No. 50-253-944