Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine IL-1 beta Recombinant Protein

Catalog No. ENRBOIL1BI Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENRBOIL1BI 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENRBOIL1BI Supplier Invitrogen™ Supplier No. RBOIL1BI
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For dilution, use a buffer containing carrier protein, e.g. PBS with 1% BSA or a neutral media containing 10% FCS and freeze in aliquots at -70°C. Activity: The biological activity of recombinant bovine IL-1-beta was determined by measuring the proliferation of mouse D10.G4.1 cells (Kaye J. et al. (1984) J. Immunol. 133:1339). The ED50 (or 1 unit of activity) for this effect in our laboratories typically falls into the range of 14.1 - 21.1 pg/mL. Expression: Mature bovine IL-1-beta was expressed in Pichia pastoris, KM71H. GenBank™ Accession # P09428, amino acids 114 - 266. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: *EAEAAPVQSI. *The underlined sequence is due to incomplete cleavage of the alpha-factor secretory signal sequence by the Ste 13 protease. Reconstitute with 0.5 mL of distilled water.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09428
For Use With (Application) Bioactivity, Immunoprecipitation, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 281251
Molecular Weight (g/mol) 17.7 kDa
Name Bovine IL-1 beta
Purification Method Purified
Quantity 20 μg
Source Yeast
Storage Requirements 4°C
Regulatory Status RUO
Gene Alias catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Common Name IL-1 beta
Gene Symbol IL1B
Biological Activity ED50 = 106-159 pg/mL; determined by the dose-dependent proliferation of mouse D10.G4.1 cells.
Product Type Protein
Conjugate Unconjugated
Species Cattle/Bovine
Recombinant Recombinant
Sequence Bovine IL-1 beta, amino acids 114-266 (Accession # P09428)
Content And Storage 4° C
Activity ED50 = 106-159 pg/mL; determined by the dose-dependent proliferation of mouse D10.G4.1 cells.
Expression System P. pastoris
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >95% by SDS-PAGE analysis
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.