Learn More
Kingfisher Biotech Inc Bovine IL-13 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0002B005
Encompass
The Bovine IL-13 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IL-13 applications are for cell culture, ELISA standard, and Western Blot Control. The Bovine IL-13 yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IL-13 Specifications: (Molecular Weight: 12.6 kda) (Amino Acid Sequence: SPVPSATALK ELIEELVNIT QNQKVPLCNG SMVWSLNLTS SMYCAALDSL ISISNCSVIQ RTKRMLNALC PHKPSAKQVS SEYVRDTKIE VAQFLKDLLR HSRIVFRNER FN (112)) (Gene ID: 281247). For research use only. Made in the USA
Catalog No. 50-241-3395