Learn More
Kingfisher Biotech Inc Bovine IL-17A Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0056B025
Encompass
The Bovine IL-17A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IL-17A applications are for cell culture, ELISA standard, and Western Blot Control. The Bovine IL-17A yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IL-17A Specifications: (Molecular Weight: 15.0 kDa) (Amino Acid Sequence: (KAGVIIPQSPGCPPTEDKNFPQHVRVNLNIVNRSTNSRRPTDYHKRSTSPWTLHRNEDPERYPSVIWEAKCSHSGCINAEGKVDHHMNSVTIQQEILVLRRESQHCPHSFRLEKMLVAVGCTCVTPIVRHLA) (Gene ID: 282863). For research use only. Made in the USA
Catalog No. 50-241-3385