Learn More
Raybiotech Inc Bovine IL-17A ELISA Kit
Supplier:  Raybiotech Inc ELBIL17A1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Bovine IL-17A
Catalog No. 50-194-8864