Learn More
Kingfisher Biotech Inc Bovine IL-18 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0304B025
Encompass
The Bovine IL-18 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IL-18 applications are for cell culture, ELISA standard, and Western Blot Control. The Bovine IL-18 yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IL-18 Specifications: (Molecular Weight: 18.1 kDa) (Amino Acid Sequence: HFGKLEPKLS IIRNLNDQVL FINQGNQPVF EDMPDSDCSD NAPQTIFIIY MYKDSLTRGL AVTISVQCKK MSTLSCENKI VSFKEMNPPD NIDNEESDII FFQRSVPGHD DKIQFESSLY EGYFLACKKE NDLFKLILKK QDDNRDKSVM FTVQNQN (157)) (Gene ID: 281249). For research use only. Made in the USA
Catalog No. 50-241-3262