Learn More
Raybiotech Inc Bovine IL-2 ELISA Kit
Supplier:  Raybiotech Inc ELBIL21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Detects bovine IL-2 in this Sandwich ELISA. It shows less than 0.2% cross-reactivity with the recombinant human IL-2, recombinant mouse IL-2, recombinant rat IL-2, recombinant cotton rat IL-2, recombinant canine IL-2, recombinant equine IL-2, recombinant feline IL-2.
Catalog No. 50-194-8867