Learn More
Raybiotech Inc Bovine IL-21 ELISA Kit
Supplier:  Raybiotech Inc ELBIL211
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.41 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: bovine IFN-alpha, IFN-gamma, IL-13, IL-1 alpha, IL-1F5, IP-10, MIG, MIP-1beta, TNF-alpha.
Catalog No. 50-194-8868