Learn More
Raybiotech Inc Bovine IL-4 ELISA Kit
Supplier:  Raybiotech Inc ELBIL41
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 8 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Bovine IL-4
Catalog No. 50-194-8869